p-Xylylenediamine

Supplier: ABCR GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1,4-Bis(aminomethyl)benzene1,4-Phenylenedimethanamine

ABCRAB143306.0025EA 0 GBP
ABCRAB143306.0025
p-Xylylenediamine
p-Xylylenediamine
Formula: C₈H₁₂N₂
MW: 136.20 g/mol
MDL Number: MFCD00009821
CAS Number: 539-48-0
EINECS: 208-719-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR