4-Acetyl-2-chloropyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-(2-chloropyridin-4-yl)ethan-1-one, 1-(2-chloropyridine-4-yl)ethanone

AS22752G1 AS22752G25 AS22752G5
ACTIAS22752G1EA 0 GBP
ACTIAS22752G1 ACTIAS22752G25 ACTIAS22752G5
4-Acetyl-2-chloropyridine
4-Acetyl-2-chloropyridine
Formula: C₇H₆ClNO
MW: 155.58 g/mol
MDL Number: MFCD07699406
CAS Number: 23794-15-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR