1-Boc-3-methylpiperazine
Supplier: ACTIVATE SCIENTIFIC GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
tert-Butyl 3-methylpiperazine-1-carboxylate, 3-Methylpiperazine, N1-BOC protected
AS20453G25
AS20453G5
ACTIAS20453G25EA
66
GBP
ACTIAS20453G25
ACTIAS20453G5
1-Boc-3-methylpiperazine
1-Boc-3-methylpiperazine
Formula:
C₁₀H₂₀N₂O₂ MW: 200.28 g/mol |
MDL Number:
MFCD03001706 CAS Number: 120737-59-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...