Trimesic acid triethyl ester, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Triethyl benzene-1,3,5-tricarboxylateTriethyl 1,3,5-benzenetricarboxylate

SIAL444693-25GEA 205 GBP
SIAL444693-25G
Trimesic acid triethyl ester, Sigma-Aldrich®
Trimesic acid triethyl ester
Formula: C₁₅H₁₈O₆
MW: 294.31 g/mol
MDL Number: MFCD00236389
CAS Number: 4105-92-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR