Dipotassium oxalate 300 g/l in aqueous solution
Supplier: PROLABO CMP
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
Potassiumoxalate
, Potassium ethanedioate
, Oxalic acid potassium salt
PRLS87203.290EA
0
GBP
PRLS87203.290
Dipotassium oxalate 300 g/l in aqueous solution
di-Potassium oxalate
Formula:
C₂K₂O₄ MW: 166.22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00012464 CAS Number: 583-52-8 EINECS: 209-506-8 Merck Index: 12,07820 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...