2,6-Dichlorobenzoxazole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21735G100 AS21735G25 AS21735G5
ACTIAS21735G100EA 76 GBP
ACTIAS21735G100 ACTIAS21735G25 ACTIAS21735G5
2,6-Dichlorobenzoxazole
2,6-Dichlorobenzoxazole
Formula: C₇H₃Cl₂NO
MDL Number: MFCD07368635
CAS Number: 3621-82-7
EINECS: 222-818-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR