4-((4,5-Dihydrothiazol-2-yl)carbamoyl)phenylboronic acid

Supplier: COMBI-BLOCKS INC.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (4-((4,5-Dihydrothiazol-2-yl)carbamoyl)phenyl)boronic acid

BB-3044-1G BB-3044-5G
COBBBB-3044-1GEA 0 GBP
COBBBB-3044-1G COBBBB-3044-5G
4-((4,5-Dihydrothiazol-2-yl)carbamoyl)phenylboronic acid
4-((4,5-Dihydrothiazol-2-yl)carbamoyl)phenylboronic acid
Formula: C₁₀H₁₁BN₂O₃S
MW: 250.0800 g/mol
MDL Number: MFCD06659881
CAS Number: 850568-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR