2-Amino-4'-bromoacetophenone hydrochloride, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Bromophenacylamine hydrochloride

415340-5G 415340-1G
SIAL415340-5GEA 53.6 GBP
SIAL415340-5G SIAL415340-1G
2-Amino-4'-bromoacetophenone hydrochloride, Sigma-Aldrich®
2-Amino-4'-bromoacetophenone hydrochloride
Formula: C₈H₉BrClNO
MW: 250.53 g/mol
MDL Number: MFCD00051998
CAS Number: 5467-72-1
EINECS: 226-778-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR