3,5-Dimethoxyphenylboronic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3,5-Dimethoxybenzeneboronic acid

AS23252G25 AS23252G5
ACTIAS23252G25EA 58 GBP
ACTIAS23252G25 ACTIAS23252G5
3,5-Dimethoxyphenylboronic acid
3,5-Dimethoxyphenylboronic acid
Formula: C₈H₁₁BO₄
MW: 181.98 g/mol
MDL Number: MFCD03095127
CAS Number: 192182-54-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR