Potassium-4-(methoxycarbonyl)phenyltrifluoroborate
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Potassium trifluoro(4-(methoxycarbonyl)phenyl)borate
TB-3208-1G
TB-3208-25G
TB-3208-5G
COBBTB-3208-1GEA
0
GBP
COBBTB-3208-1G
COBBTB-3208-25G
COBBTB-3208-5G
Potassium-4-(methoxycarbonyl)phenyltrifluoroborate
Potassium 4-(methoxycarbonyl)phenyltrifluoroborate
Formula:
C₈H₇BF₃KO₂ MW: 242.05 g/mol |
MDL Number:
MFCD04115741 CAS Number: 705254-34-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...