Yttrium, powder, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ100

261327-5G
SIAL261327-5GEA 159 GBP
SIAL261327-5G
Yttrium, powder, Sigma-Aldrich®
Yttrium
Formula: Y
MDL Number: MFCD00011468
CAS Number: 7440-65-5
EINECS: 231-174-8
UN: 3089
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR