3',4'-Dimethylacetophenone, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ100

137235-25G
SIAL137235-25GEA 133 GBP
SIAL137235-25G
3',4'-Dimethylacetophenone, Sigma-Aldrich®
3',4'-Dimethylacetophenone
Formula: C₁₀H₁₂O
Boiling Pt: 243 °C (1013 hPa)
Melting Pt: –3 °C
Density: 1.0185 g/cm³ (0 °C)
MDL Number: MFCD00008743
CAS Number: 3637-01-2
EINECS: 222-859-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR