α,α,α-Trifluoro-o-cresol

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

35352855-5G
MOLE35352855-5GEA 0 GBP
MOLE35352855-5G
α,α,α-Trifluoro-o-cresol
α,α,α-Trifluoro-o-cresol
Formula: C₇H₅F₃O
Boiling Pt: 147 °C (1013 hPa)
Melting Pt: 46 °C
Density: >1 g/cm³ (20 °C)
MDL Number: MFCD00002222
CAS Number: 444-30-4
EINECS: 207-148-7
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR