Tyramine hydrochloride

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 2-(4-Hydroxyphenyl)ethylammonium chloride, 4-(2-Aminoethyl)phenol hydrochloride, 4-Hydroxyphenethylammonium chloride

13456061-25G
MOLE13456061-25GEA 0 GBP
MOLE13456061-25G
Tyramine hydrochloride
Tyramine hydrochloride
Formula: C₈H₁₂ClNO
MW: 173.64 g/mol
Melting Pt: 270…272 °C
MDL Number: MFCD00012901
CAS Number: 60-19-5
EINECS: 200-462-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR