Masterflex® Male Rotating Luer Lock Plug Fitting, Avantor®
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MFLX40320-00EA 14.6 GBP
MFLX40320-00 MFLX40320-01 MFLX40320-02 MFLX40320-03 MFLX40320-04 MFLX40320-05 MFLX40320-06 MFLX40320-07 MFLX40320-08 MFLX40320-09 MFLX40320-10 MFLX40320-11 MFLX40320-12 MFLX40320-13 MFLX40320-14 MFLX40320-15 MFLX40320-16 MFLX40320-17 MFLX40320-18 MFLX40320-19 MFLX40320-20
Masterflex® Male Rotating Luer Lock Plug Fitting, Avantor®
Fittings and Connectors
Easily transition to a luer fitting.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Determine which connector type is needed for your application by contacting free technical application assistance
These fittings are manufactured in an ISO Class 7 cleanroom at an ISO 9001 facility; utilizing an ISO 13485 quality system. There is full lot traceability and tight dimensional tolerances forMore Product Information