Masterflex® Transfer Tubing, C-Flex® ULTRA, Avantor®
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
06432-45 06432-49
MFLX06432-41EA 150 GBP
MFLX06432-41 MFLX06432-42 MFLX06432-47 MFLX06432-48 MFLX06432-43 MFLX06432-45 MFLX06432-49
Masterflex® Transfer Tubing, C-Flex® ULTRA, Avantor®
Tubing Transfer Tubing
Excellent resistance to concentrated acids and alkalis.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
Applications:Bioprocess, biopharmaceutical, single-use assemblies, process filling, sampling.
Characteristics: Animal-derived component-free (ADCF). Sealable and weldable. Noncytotoxic and nonpyrogenic;More Product Information