Specimen clamps for rotary microtomes
Supplier: LEICA MICROSYSTEMS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
14050237999 14050238002
LEIC14050237999EA 702 GBP
LEIC14050237999 LEIC14050238002
Specimen clamps for rotary microtomes
Clamps General Purpose Clamps
These clamps can be integrated into the fine directional, directional, or non-directional specimen holder fixtures.