XPert® TXE™ Equipment Enclosures
Supplier: LABCONCO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
3892020 3892021
LBCP3892020EA 11250 GBP
LBCP3892020 LBCP3892021
XPert® TXE™ Equipment Enclosures
Fume Hoods and Enclosures Balances Enclosures
XPert TXE Equipment Enclosures are designed to address the needs of pharmaceutical research and quality control laboratories.
These vented equipment enclosures may be ducted to the outside or connected to the filtermate portable exhauster for HEPA and/or carbon filtered exhaust back into the laboratory. Specialized base stands are also available (sold separately).
It provides a safe, ventilated space for keeping powder-generating apparatus contained. Its tall 1066,8 mm interior height accommodates equipment such as pill presses, grinders, crushers and sieves. HEPA-filtered air isMore Product Information