tert-Butyl acrylate 99% stabilised

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger


Stabilisation: Stabilised with 4-methoxyphenol (MEHQ) 15 ppm

L17169.AE L17169.AP L17169.AU
L17169.AEEA 24.1 GBP
L17169.AE L17169.AP L17169.AU
tert-Butyl acrylate 99% stabilised
tert-Butyl acrylate

Stab. with 15ppm 4-methoxyphenol


Purity: 99% CAS No.: 1663-39-4 Molecular Formula: C7H12O2 Formula Weight: 128.17 Flash Point: 11°(52°F) Refractive Index: 1.4110 Melting Point: ca -69° Boiling Point: 121° Beilstein Reference: 1742329 UN No.: UN3272 MDL No.: MFCD00008809

Formula: C₇H₁₂O₂
Boiling Pt: 121 °C
Melting Pt: ∼–69 °C
Density: 0.883
Flash Pt: 11 °C (52 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008809
CAS Number: 1663-39-4
EINECS: 216-768-7
UN: 3272
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR