Settle plates, gamma-irridiated, triple-bagged, ICR (isolator, cleanroom)
Supplier: MERCK LIFE SCIENCE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1460690120 1460010020 1460010120 1460050120 1460050020 1460500020 1460690020 1460760120
HEIP1460690120EA 654 GBP
HEIP1460690120 HEIP1460010020 HEIP1460010120 HEIP1460050120 HEIP1460050020 HEIP1460500020 HEIP1460690020 HEIP1460760120
Settle plates, gamma-irridiated, triple-bagged, ICR (isolator, cleanroom)
Media for Microbiology Prepared Media for Microbiology
A wide range of ready-to-use culture media for the environmental monitoring of surfaces as well as for the active and passive air sampling in clean rooms and isolators. ICRplus settle plates for efficient anaerobic incubation in isolators and clean rooms.
  • Gamma-irradiated
  • Triple-bagged
Order Now
SPECIFICATIONS
Environmentally Preferable