(S)-3-Amino-5-hexynoic acid hydrochloride 95%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

H52010.MD H52010.03
H52010.MDEA 162 GBP
H52010.MD H52010.03
(S)-3-Amino-5-hexynoic acid hydrochloride 95%
(S)-3-Amino-5-hexynoic acid hydrochloride

Purity: 95% CAS No.: 270596-46-8 Molecular Formula: C6H9NO2.HCl Formula Weight: 163.60 MDL No.: MFCD01861006

Formula: C₆H₉NO₂·HCl
Storage Temperature: Ambient
MDL Number: MFCD01861006
CAS Number: 270596-46-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR