1-Fluoro-3,5-dimethyl-2-nitrobenzene 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

H27762.03
H27762.03EA 43.3 GBP
H27762.03
1-Fluoro-3,5-dimethyl-2-nitrobenzene 98%
1-Fluoro-3,5-dimethyl-2-nitrobenzene

Purity: 98% CAS No.: 315-13-9 Molecular Formula: C8H8FNO2 Formula Weight: 169.16 Melting Point: 54-57° MDL No.: MFCD00052367

Formula: C₈H₈FNO₂
Melting Pt: 54…57 °C
Storage Temperature: Ambient
MDL Number: MFCD00052367
CAS Number: 315-13-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR