3-Phenoxybenzyl chloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

018948-1G 018948-5G 018948-25G
FLUO018948-1GEA 0 GBP
FLUO018948-1G FLUO018948-5G FLUO018948-25G
3-Phenoxybenzyl chloride
3-Phenoxybenzyl chloride
Formula: C₁₃H₁₁ClO
Boiling Pt: 123 °C (0.3 mmHg)
Density: 1.16 g/cm³ (20 °C)
MDL Number: MFCD00040866
CAS Number: 53874-66-1
EINECS: 258-831-1
UN: 3265
ADR: 8,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR