1-Fluoronaphthalene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

002697-25G 002697-100G 002697-500G
FLUO002697-25GEA 0 GBP
FLUO002697-25G FLUO002697-100G FLUO002697-500G
1-Fluoronaphthalene
1-Fluoronaphthalene
Formula: C₁₀H₇F
Boiling Pt: 215 °C (1013 hPa)
Melting Pt: –12.5 °C
Density: 1,135 g/cm³ (0 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003873
CAS Number: 321-38-0
EINECS: 206-287-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR