Robust Temperature Data Logger, EBI 12-T100
Supplier: EBRO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1340-6600 1340-6600-EX 1340-6600-0100
EBRO1340-6600EA 2170 GBP
EBRO1340-6600 EBRO1340-6600-EX EBRO1340-6600-0100
Robust Temperature Data Logger, EBI 12-T100
Data Loggers
Discover the EBI 12-T100 from ebro: a robust temperature data logger, ideal for independent routine checks in medicine, pharmaceuticals and production. Sterilisable and with a large measuring range for the optimal quality assurance.
  • Wide measuring range, –90 to +150 °C
  • High-quality PT1000 sensor (class A)
  • Robust IP 68 protection class: waterproof and vapour-tight
  • Supplied with calibration certificate
  • Sterilisable data logger
The EBI 12-T100 from ebro is a high-performance temperature data logger. Its impressive measuring range of –90 to +150 °C andMore Product Information