Anti-AANAT Rabbit Polyclonal Antibody

Supplier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-3914R
BOSSBS-3914REA 680 GBP
BOSSBS-3914R
Anti-AANAT Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: AANAT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR