Anti-eNOS Ser615 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Supplier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-13073R-A680
BOSSBS-13073R-A680EA 860 GBP
BOSSBS-13073R-A680
Anti-eNOS Ser615 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
eNOS is a calcium/calmodulin dependent enzyme which undergoes several post translational modifications, including acylation with myristate and palmitate, and phosphorylation on numerous residues. As with the other members of the NOS family, eNOS derives the diffusible multifunctional second messenger NO from L arginine through a series of reactions in which L citrulline is an intermediate. eNOS plays an important role in controlling vascular tone, platelet aggregation, and cardiac myocyte function.

Type: Primary
Antigen: eNOS Ser615
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR