Anti-KRT7 Mouse Monoclonal Antibody (Biotin) [clone: KRT7/760]
Supplier: Biotium
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BNCB0760-500 BNCB0760-100
BNCB0760-500EA 533 GBP
BNCB0760-500 BNCB0760-100
Anti-KRT7 Mouse Monoclonal Antibody (Biotin) [clone: KRT7/760]
Antibodies
This antibody recognizes an intermediate filament protein (IFP) of 55 kDa, which is identified as cytokeratin 7. This MAb is highly specific to cytokeratin 7 and shows no cross-reaction with other IFPs. Cytokeratin 7 is a basic cytokeratin, which is found in most glandular and transitional epithelia but not in the stratified squamous epithelia. Keratin 7 is expressed in the epithelial cells of ovary, lung, and breast but not of colon, prostate, or gastrointestinal tract. This MAb is highly useful in distinguishing ovarian carcinomas (keratin 7 ) from colon carcinomas (keratin 7-).