n-Octyl-β-D-maltoside

Supplier: BIOMOL RESEARCH LABORATORIES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

54560-1 54560-2
BMOL54560-1EA 0 GBP
BMOL54560-1 BMOL54560-2
n-Octyl-β-D-maltoside
n-Octyl-β-D-maltoside
Formula: C₂₀H₃₈O₁₁
CAS Number: 82494-08-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR