4-(Pentafluorothio)benzoyl chloride 97+%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

PC0813-1G PC0813-5G PC0813-25G
APOSPC0813-1GEA 210 GBP
APOSPC0813-1G APOSPC0813-5G APOSPC0813-25G
4-(Pentafluorothio)benzoyl chloride 97+%
4-(Pentafluorothio)benzoyl chloride
Formula: C₇H₄ClF₅OS
Storage Temperature: Ambient
MDL Number: MFCD03788523
CAS Number: 197384-98-8
UN: 1760
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR