1-Isopropyl-1H-pyrazol-4-amine 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR962707-10G OR962707-1G OR962707-5G
APOSOR962707-10GEA 254 GBP
APOSOR962707-10G APOSOR962707-1G APOSOR962707-5G
1-Isopropyl-1H-pyrazol-4-amine 98%
1-Isopropyl-1H-pyrazol-4-amine
Formula: C₆H₁₁N₃
Storage Temperature: Ambient
MDL Number: MFCD08700622
CAS Number: 97421-16-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR