Ethyl tetrahydropyran-4-carboxylate 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR952913-5G
APOSOR952913-5GEA 52.5 GBP
APOSOR952913-5G
Ethyl tetrahydropyran-4-carboxylate 98%
Ethyl tetrahydropyran-4-carboxylate
Formula: C₈H₁₄O₃
Storage Temperature: Ambient
MDL Number: MFCD08702763
CAS Number: 96835-17-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR