4-Chloro-6-methyl-1H-indazole ≥98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR950400-1G OR950400-250MG
APOSOR950400-1GEA 451 GBP
APOSOR950400-1G APOSOR950400-250MG
4-Chloro-6-methyl-1H-indazole ≥98%
4-Chloro-6-methyl-1H-indazole
Formula: C₈H₇ClN₂
Storage Temperature: Ambient
MDL Number: MFCD07781503
CAS Number: 885521-74-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR