1-(3-Pyridin-3-ylisoxazol-5-yl)methanamine 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR930141-1G OR930141-250MG
APOSOR930141-1GEA 655 GBP
APOSOR930141-1G APOSOR930141-250MG
1-(3-Pyridin-3-ylisoxazol-5-yl)methanamine 95%
1-(3-Pyridin-3-ylisoxazol-5-yl)methanamine
Formula: C₉H₉N₃O
Storage Temperature: Refrigerator
MDL Number: MFCD08059814
CAS Number: 543713-43-5
UN: 3259
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR