4,6-Dihydrothieno[3,4-b]thiophene-2-carboxylic acid 96%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR54533-250MG
APOSOR54533-250MGEA 20 GBP
APOSOR54533-250MG
4,6-Dihydrothieno[3,4-b]thiophene-2-carboxylic acid 96%
4,6-Dihydrothieno[3,4-b]thiophene-2-carboxylic acid
Formula: C₇H₆O₂S₂
MW: 186.24 g/mol
Melting Pt: 240 °C
Storage Temperature: Ambient
MDL Number: MFCD20642620
CAS Number: 7712-05-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR