3-Chloro-5-iodobenzyl bromide ≥95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR400246-250MG OR400246-1G
APOSOR400246-250MGEA 221 GBP
APOSOR400246-250MG APOSOR400246-1G
3-Chloro-5-iodobenzyl bromide ≥95%
3-Chloro-5-iodobenzyl bromide
Formula: C₇H₅BrClI
MDL Number: MFCD18393369
CAS Number: 912343-41-0
UN: 3261

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR