3-(Dimethylamino)-1-phenyl-2-buten-1-one 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR33117-1G OR33117-5G
APOSOR33117-1GEA 195 GBP
APOSOR33117-1G APOSOR33117-5G
3-(Dimethylamino)-1-phenyl-2-buten-1-one 95%
3-(Dimethylamino)-1-phenyl-2-buten-1-one
Formula: C₁₂H₁₅NO
Storage Temperature: Ambient
MDL Number: MFCD08056660
CAS Number: 34523-87-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR