N-Methyl-2-(piperidin-4-yl)benzamide hydrochloride 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR321246-100MG
APOSOR321246-100MGEA 313 GBP
APOSOR321246-100MG
N-Methyl-2-(piperidin-4-yl)benzamide hydrochloride 97%
N-Methyl-2-(piperidin-4-yl)benzamide hydrochloride
Formula: C₁₃H₁₈N₂O·ClH
MW: 254.756 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD30099248
CAS Number: 2244085-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR