Dimethyl-1H-pyrazole-3,5-dicarboxylate

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR12087-1G
APOSOR12087-1GEA 16.8 GBP
APOSOR12087-1G
Dimethyl-1H-pyrazole-3,5-dicarboxylate
Dimethyl-1H-pyrazole-3,5-dicarboxylate
Formula: C₇H₈N₂O₄
Melting Pt: 158…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00975602
CAS Number: 4077-76-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR