1,4-Diisocyanatobutane 97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

314380010 314380050 314380250
ACRO314380010EA 55.9 GBP
ACRO314380010 ACRO314380050 ACRO314380250
1,4-Diisocyanatobutane 97%
1,4-Diisocyanatobutane
Formula: C₆H₈N₂O₂
Boiling Pt: 102…104 °C (14 mmHg)
Melting Pt: 185…187 °C
Density: 1.1 g/cm³
Flash Pt: 106 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00075559
CAS Number: 4538-37-8
UN: 2206
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4510 to 1.4530 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR