Anti-MSN Mouse Monoclonal Antibody [clone: MSN491]

Supplier: Abnova
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

MAB12138
ABNOMAB12138EA 517 GBP
ABNOMAB12138
Anti-MSN Mouse Monoclonal Antibody [clone: MSN491]
Antibodies
Mouse monoclonal antibody raised against recombinant human MSN.

Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: MSN
Clonality: Monoclonal
Clone: MSN491
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

SPECIFICATIONS

Antigen symbol MSN
Antigen name Moesin
Environmentally Preferable
Antigen synonyms Membrane-organizing Extension Spike Protein
Antibody type Primary
Clonality Monoclonal
Conjugation Unconjugated
Clone MSN491
Reactivity
Host Mouse
Gene ID 4478
Isotype IgG1 kappa
Western blot Yes
ImmunoChemistry Yes
Immunogen Recombinant protein corresponding to human MSN.
Format Liquid
Purification Protein A/G purification
Cross adsorption No
Storage buffer PBS (0,05% BSA, 0,05% sodium azide).
Storage temperature Store at 4 °C. Aliquot to avoid repeated freezing and thawing.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR