Citral cis- and trans-mixture 95%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

A16169.18 A16169.22 A16169.36
A16169.18EA 19.7 GBP
A16169.18 A16169.22 A16169.36
Citral cis- and trans-mixture 95%
Citral

Purity: 95% CAS No.: 5392-40-5 Molecular Formula: C10H16O Formula Weight: 152.24 Storage Temperature: 2°C to 8°C Flash Point: 98°(208°F) Refractive Index: 1.4880 Melting Point: -10° Boiling Point: 228-229° Beilstein Reference: 1721871 Merck Reference: 14,2322 MDL No.: MFCD00006997

Formula: C₁₀H₁₆O
Boiling Pt: 228…229 °C
Melting Pt: < –10 °C
Density: 0.888
Flash Pt: 98 °C (208 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00006997
CAS Number: 5392-40-5
EINECS: 226-394-6
Merck Index: 14,2322

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR