Tetraheptylammonium bromide, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning


M-Clarity™ quality level = MQ100

87296-10G-F 87296-50G-F
87296-10G-FEA 197 GBP
87296-10G-F 87296-50G-F
Tetraheptylammonium bromide, Supelco®
Tetraheptylammonium bromide
Formula: C₂₈H₆₀BrN
Melting Pt: 96 °C
MDL Number: MFCD00011861
CAS Number: 4368-51-8
EINECS: 224-459-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b02cb266f9a1f8dd8d5d5e4f11ad8c69e47b90b&vid=7691c2e7f3a8e02e879853712adec7b46378801d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR