VWR®, Gardner colour standards
Supplier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
84815.260EA 203 GBP
84815.260 84816.260 84817.260 84818.260 84819.260 84820.260 84821.260 84822.260
VWR®, Gardner colour standards
Standards Colour Reference and Spectroscopy Standards
Ready to use colour solutions that are prepared gravimetrically on a weight/weight basis that are designed for grading the colour of similarly coloured liquids, such as resins, varnishes, lacquers, drying oils, fatty acids, lecithin's, sunflower and linseed oil.
  • Use either as calibration and/or quality control standards
  • Designed specifically for use in ASTM analytical methods, and produced in accordance with ASTM D1500, D6045, D1209
  • Traceable product certified by an independent laboratory
  • Supplied in high quality, tamper-evident bottles