Lithium nitrate, anhydrous ≥99.999% (metals basis)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger


Packaged under argon.

44456.09 44456.18 44456.30
44456.09EA 66.2 GBP
44456.09 44456.18 44456.30
Lithium nitrate, anhydrous ≥99.999% (metals basis)
Lithium nitrate

Purity: 99.999% CAS No.: 7790-69-4 Molecular Formula: LiNO3 Formula Weight: 68.95 Physical Form: Crystalline/Lump, Packaged under argon Density: 2.38 Appearance: White Odour: Odourless Melting Point: 264° Boiling Point: 600° dec. Merck Reference: 14,5536 UN No.: 2722 MDL No.: MFCD00011094

Formula: LiNO₃
Boiling Pt: 600 °C (dec.)
Melting Pt: 264 °C
Density: 2.38
Storage Temperature: Ambient
MDL Number: MFCD00011094
CAS Number: 7790-69-4
EINECS: 232-218-9
UN: 2722
ADR: 5.1,III
Merck Index: 14,5536

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR