Giemsa's stain modified solution, Sigma-Aldrich®

Supplier: SIGMA ALDRICH MICROSCOPY
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger


M-Clarity™ quality level = MQ200

32884-1L 32884-250ML
32884-1LEA 106 GBP
32884-1L 32884-250ML
Giemsa's stain modified solution, Sigma-Aldrich®
Giemsa's stain

Modified Giemsa stain may be used for the identification of each human chromosome by differentials staining.


Modified Giemsa stain means when the pH of the Giemsa stain is changed from the usual 6.8 to 9.0.

Density: 0.99 g/cm³ (20 °C)
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00081642
CAS Number: 51811-82-6
EINECS: 257-438-2
UN: 1230
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR