Funnels, powder, Nalgene®
Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
4252-0065 4252-0080 4252-0150 4252-0100
221-0090EA 106 GBP
221-0090 221-0091 221-0093 221-0092
Funnels, powder, Nalgene®
Funnels Powder Funnels
PPCO, translucent.
  • Very good chemical resistance
  • Outer ribs prevent air from being trapped
  • Autoclavable
For transferring powdery materials. Parallel stem prevents blockages.
This shatter-proof alternative to glass is chemical resistant to a wide range of lab chemicals and reagents and can make a valuble contribution to lab safety programs.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification IRECS; ISO 14001 (Certificate Registration No. EMS 752603)