Ethyl eosin ≥95% (dye content), certified by the Biological Stain Commission, Sigma-Aldrich®

Supplier: SIGMA ALDRICH MICROSCOPY
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ200

199540-25G 199540-5G
199540-25GEA 314 GBP
199540-25G 199540-5G
Ethyl eosin ≥95% (dye content), certified by the Biological Stain Commission, Sigma-Aldrich®
Ethyl eosin

Certified for use in the Stoval and Black method for demonstrating Negri bodies in the central nervous system of rabid animals.

Formula: C₂₂H₁₁Br₄KO₅
Storage Temperature: Ambient
MDL Number: MFCD00005039
CAS Number: 6359-05-3

Order Now

Specification Test Results

λmax 532 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR