VWR® VP 820, Diaphragm Vacuum Pump
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
181-0071EA 1810 GBP
181-0071 181-0072 181-0073
VWR® VP 820, Diaphragm Vacuum Pump
Pumps Vacuum Diaphragm Pumps
Dual-head, dry running diaphragm pump for a wide range of laboratory applications. It pumps, compresses and evacuates without affecting media, i.e. media remains uncontaminated. The pump includes a diaphragm with patented tension optimisation, which allows the pump to be smaller prolonging the service life of the diaphragm.
  • 100% oil-free pumping, so media cannot be contaminated when pumping and evacuating
  • For medium aggressive gases and vapours
  • Maintenance-free, quiet running
* Flow rate at atmospheric pressure
Delivery information: IncludesMore Product Information