Chloropentaamminerhodium(III) chloride (≥34.5% Rh)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

10519.02 10519.03 10519.06
10519.02EA 409 GBP
10519.02 10519.03 10519.06
Chloropentaamminerhodium(III) chloride (≥34.5% Rh)
Chloropentaamminerhodium(III) chloride

CAS No.: 13820-95-6 Molecular Formula: [RhCl(NH3)5]Cl2 Formula Weight: 294.42 Physical Form: Micro Crystals Sensitivity: Hygroscopic Melting Point: dec. MDL No.: MFCD00135665

Formula: [RhCl(NH₃)₅]Cl₂
Melting Pt: Dec.
Storage Temperature: Ambient
MDL Number: MFCD00135665
CAS Number: 13820-95-6
EINECS: 237-505-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR